Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DSTN Full-length ORF (AAH09477.1, 1 a.a. - 165 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00011034P02L

 View more versions of this product

Catalog No. 89-942-486


Add to cart

Description

Description

The product of this gene belongs to the actin-binding proteins ADF family. This family of proteins is responsible for enhancing the turnover rate of actin in vivo. This gene encodes the actin depolymerizing protein that severs actin filaments (F-actin) and binds to actin monomers (G-actin). Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]

Sequence: MASGVQVADEVCRIFYDMKVRKCSTPEEIKKRKKAVIFCLSADKKCIIVEEGKEILVGDVGVTITDPFKHFVGMLPEKDCRYALYDASFETKESRKEELMFFLWAPELAPLKSKMIYASSKDAIKKKFQGIKHECQANGPEDLNRACIAEKLGGSLIVAFEGCPV
Specifications

Specifications

AAH09477.1
Liquid
11034
DSTN (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MASGVQVADEVCRIFYDMKVRKCSTPEEIKKRKKAVIFCLSADKKCIIVEEGKEILVGDVGVTITDPFKHFVGMLPEKDCRYALYDASFETKESRKEELMFFLWAPELAPLKSKMIYASSKDAIKKKFQGIKHECQANGPEDLNRACIAEKLGGSLIVAFEGCPV
RUO
DSTN
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
44.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ACTDP|ADF|bA462D18.2
DSTN
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit