Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DTNB Full-length ORF (AAH16655.1, 1 a.a. - 560 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00001838P01

Catalog No. 89-942-496


Add to cart

Description

Description

This gene encodes dystrobrevin beta, a component of the dystrophin-associated protein complex (DPC). The DPC consists of dystrophin and several integral and peripheral membrane proteins, including dystroglycans, sarcoglycans, syntrophins and dystrobrevin alpha and beta. The DPC localizes to the sarcolemma and its disruption is associated with various forms of muscular dystrophy. Dystrobrevin beta is thought to interact with syntrophin and the DP71 short form of dystrophin. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Sequence: MIEESGNKRKTMAEKRQLFIEMRAQNFDVIRLSTYRTACKLRFVQKRCNLHLVDIWNMIEAFRDNGLNTLDHTTEISVSRLETVISSIYYQLNKRLPSTHQISVEQSISLLLNFMIAAYDSEGRGKLTVFSVKAMLATMCGGKMLDKLRYVFSQMSDSNGLMIFSKFDQFLKEVLKLPTAVFEGPSFGYTEHSVRTCFPQQRKIMLNMFLDTMMADPPPQCLVWLPLMHRLAHVENVFHPVECSYCRCESMMGFRYRCQQCHNYQLCQNCFWRGHAGGPHSNQHQMKEHSSWKSPAKKLSHAISKSLGCVPTREPPHPVFPEQPEKPLDLAHIVPPRPLTNMNDTMVSHMSSGVPTPTKSVLDSPSRLDEEHRLIARYAARLAAEAGNVTRPPTDLSFNFDANKQQRQLIAELENKNREILQEIQRLRLEHEQASQPTPEKAQQNPTLLAELRLLRQRKDELEQRMSALQESRRELMVQLEELMKLLKAQATGSPHTSPTHGGGRPMPMPVRSTSAGSTPTHCPQDSLSGVGGDVQEAFAQAEEGAEEEEEKMQNGKDRG
Specifications

Specifications

AAH16655.1
Liquid
1838
DTNB (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MIEESGNKRKTMAEKRQLFIEMRAQNFDVIRLSTYRTACKLRFVQKRCNLHLVDIWNMIEAFRDNGLNTLDHTTEISVSRLETVISSIYYQLNKRLPSTHQISVEQSISLLLNFMIAAYDSEGRGKLTVFSVKAMLATMCGGKMLDKLRYVFSQMSDSNGLMIFSKFDQFLKEVLKLPTAVFEGPSFGYTEHSVRTCFPQQRKIMLNMFLDTMMADPPPQCLVWLPLMHRLAHVENVFHPVECSYCRCESMMGFRYRCQQCHNYQLCQNCFWRGHAGGPHSNQHQMKEHSSWKSPAKKLSHAISKSLGCVPTREPPHPVFPEQPEKPLDLAHIVPPRPLTNMNDTMVSHMSSGVPTPTKSVLDSPSRLDEEHRLIARYAARLAAEAGNVTRPPTDLSFNFDANKQQRQLIAELENKNREILQEIQRLRLEHEQASQPTPEKAQQNPTLLAELRLLRQRKDELEQRMSALQESRRELMVQLEELMKLLKAQATGSPHTSPTHGGGRPMPMPVRSTSAGSTPTHCPQDSLSGVGGDVQEAFAQAEEGAEEEEEKMQNGKDRG
RUO
DTNB
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
87.23kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC17163|MGC57126
DTNB
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit