Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DTNBP1 Full-length ORF (NP_115498.2, 1 a.a. - 351 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00084062P01L

 View more versions of this product

Catalog No. 89-942-501


Add to cart

Description

Description

This gene encodes a protein that may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. A similar protein in mouse is a component of a protein complex termed biogenesis of lysosome-related organelles complex 1 (BLOC-1), and binds to alpha- and beta-dystrobrevins, which are components of the dystrophin-associated protein complex (DPC). Mutations in this gene are associated with Hermansky-Pudlak syndrome type 7. This gene may also be associated with schizophrenia. Multiple transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]

Sequence: MLETLRERLLSVQQDFTSGLKTLSDKSREAKVKSKPRTVPFLPKYSAGLELLSRYEDTWAALHRRAKDCASAGELVDSEVVMLSAHWEKKKTSLVELQEQLQQLPALIADLESMTANLTHLEASFEEVENNLLHLEDLCGQCELERCKHMQSQQLENYKKNKRKELETFKAELDAEHAQKVLEMEHTQQMKLKERQKFFEEAFQQDMEQYLSTGYLQIAERREPIGSMSSMEVNVDMLEQMDLMDISDQEALDVFLNSGGEENTVLSPALGPESSTCQNEITLQVPNPSELRAKPPSSSSTCTDSATRDISEGGESPVVQSDEEEVQVDTALATSHTDREATPDGGEDSDS
Specifications

Specifications

NP_115498.2
Liquid
84062
DTNBP1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MLETLRERLLSVQQDFTSGLKTLSDKSREAKVKSKPRTVPFLPKYSAGLELLSRYEDTWAALHRRAKDCASAGELVDSEVVMLSAHWEKKKTSLVELQEQLQQLPALIADLESMTANLTHLEASFEEVENNLLHLEDLCGQCELERCKHMQSQQLENYKKNKRKELETFKAELDAEHAQKVLEMEHTQQMKLKERQKFFEEAFQQDMEQYLSTGYLQIAERREPIGSMSSMEVNVDMLEQMDLMDISDQEALDVFLNSGGEENTVLSPALGPESSTCQNEITLQVPNPSELRAKPPSSSSTCTDSATRDISEGGESPVVQSDEEEVQVDTALATSHTDREATPDGGEDSDS
RUO
DTNBP1
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
65.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DBND|DKFZp564K192|FLJ30031|HPS7|MGC20210|My031|SDY
DTNBP1
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit