Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DUSP11 Full-length ORF (NP_003575.1, 1 a.a. - 330 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00008446P01L

 View more versions of this product

Catalog No. 89-942-540


Add to cart

Description

Description

The protein encoded by this gene is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the mitogen-activated protein (MAP) kinase superfamily (MAPK/ERK, SAPK/JNK, p38), which is associated with cellular proliferation and differentiation. Different members of the family of dual specificity phosphatases show distinct substrate specificities for various MAP kinases, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. This gene product is localized to the nucleus and binds directly to RNA and splicing factors, and thus it is suggested to participate in nuclear mRNA metabolism. [provided by RefSeq]

Sequence: MSQWHHPRSGWGRRRDFSGRSSAKKKGGNHIPERWKDYLPVGQRMPGTRFIAFKVPLQKSFEKKLAPEECFSPLDLFNKIREQNEELGLIIDLTYTQRYYKPEDLPETVPYLKIFTVGHQVPDDETIFKFKHAVNGFLKENKDNDKLIGVHCTHGLNRTGYLICRYLIDVEGVRPDDAIELFNRCRGHCLERQNYIEDLQNGPIRKNWNSSVPRSSDFEDSAHLMQPVHNKPVKQGPRYNLHQIQGHSAPRHFHTQTQSLQQSVRKFSENPHVYQRHHLPPPGPPGEDYSHRRYSWNVKPNASRAAQDRRRWYPYNYSRLSYPACWEWTQ
Specifications

Specifications

NP_003575.1
Liquid
8446
DUSP11 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSQWHHPRSGWGRRRDFSGRSSAKKKGGNHIPERWKDYLPVGQRMPGTRFIAFKVPLQKSFEKKLAPEECFSPLDLFNKIREQNEELGLIIDLTYTQRYYKPEDLPETVPYLKIFTVGHQVPDDETIFKFKHAVNGFLKENKDNDKLIGVHCTHGLNRTGYLICRYLIDVEGVRPDDAIELFNRCRGHCLERQNYIEDLQNGPIRKNWNSSVPRSSDFEDSAHLMQPVHNKPVKQGPRYNLHQIQGHSAPRHFHTQTQSLQQSVRKFSENPHVYQRHHLPPPGPPGEDYSHRRYSWNVKPNASRAAQDRRRWYPYNYSRLSYPACWEWTQ
RUO
DUSP11
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
65.3kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PIR1
DUSP11
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit