Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DUX3 Full-length ORF (NP_036280.1, 1 a.a. - 170 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00026582P01

Catalog No. 89-942-592


Add to cart

Description

Description

The human genome contains hundreds of repeats of the 3.3-kb family in regions associated with heterochromatin. The DUX gene family, including DUX3, resides within these 3.3-kb repeated elements (Beckers et al., 2001 [PubMed 11245978]). See DUX4 (MIM 606009).[supplied by OMIM]

Sequence: MALLTALDDTLPEEAQGPGRRMILLSTPSQSDALRACFERNLYPGIATKEQLAQGIDIPEPRVQIWFQNERSCQLRQHRRQSRPWPGRRDPQKGRRKRTAITGSQTALLLRAFEKDRFPGIPAREELARETGLPESRIQLWFQNRRARHWGQSGRAPTQASIRCNAAPIG
Specifications

Specifications

NP_036280.1
Liquid
26582
DUX3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MALLTALDDTLPEEAQGPGRRMILLSTPSQSDALRACFERNLYPGIATKEQLAQGIDIPEPRVQIWFQNERSCQLRQHRRQSRPWPGRRDPQKGRRKRTAITGSQTALLLRAFEKDRFPGIPAREELARETGLPESRIQLWFQNRRARHWGQSGRAPTQASIRCNAAPIG
RUO
DUX3
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
45.8kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DUX3
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit