Learn More
Abnova™ Human DYRK1B (NP_004705.1, 1 a.a. - 629 a.a.) Full-length Recombinant Protein with GST-tag expressed in baculovirus infected Sf21 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P5540
Catalog No. 89-942-633
Add to cart
Description
Description
DYRK1B is a member of the DYRK family of protein kinases. DYRK1B contains a bipartite nuclear localization signal and is found mainly in muscle and testis. The protein is proposed to be involved in the regulation of nuclear functions. Three isoforms of DYRK1B have been identified differing in the presence of two alternatively spliced exons within the catalytic domain. [provided by RefSeq]
Specifications
Specifications
NP_004705.1
Liquid
9149
DYRK1B (Human) Recombinant Protein
Glutathione sepharose chromatography
Loading 1 ug protein in SDS-PAGE
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MIRK
DYRK1B
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg(or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip 3000. Substrate: DYRKtide-F. ATP: 100 uM.
Yes
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
96kDa
Baculovirus infected insect cell (Sf21) expression system
81% by SDS-PAGE/CBB staining
5 ug
RUO
DYRK1B
Baculovirus infected insect cell (Sf21) expression system
Insect
GST