Learn More
Abnova™ Human DYRK2 (NP_003574.1, 1 a.a. - 528 a.a.) Full-length Recombinant Protein with GST-tag expressed in baculovirus infected Sf21 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P5541
Catalog No. 89-942-639
Add to cart
Description
Description
DYRK2 belongs to a family of protein kinases whose members are presumed to be involved in cellular growth and/or development. The family is defined by structural similarity of their kinase domains and their capability to autophosphorylate on tyrosine residues. DYRK2 has demonstrated tyrosine autophosphorylation and catalyzed phosphorylation of histones H3 and H2B in vitro. Two isoforms of DYRK2 have been isolated. The predominant isoform, isoform 1, lacks a 5' terminal insert. [provided by RefSeq]
Specifications
Specifications
NP_003574.1
Liquid
8445
DYRK2 (Human) Recombinant Protein
Glutathione sepharose chromatography
Loading 1 ug protein in SDS-PAGE
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ21217|FLJ21365
DYRK2
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg(or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip 3000. Substrate: DYRKtide-F. ATP: 100 uM.
Yes
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
87kDa
Baculovirus infected insect cell (Sf21) expression system
98% by SDS-PAGE/CBB staining
5 ug
RUO
DYRK2
Baculovirus infected insect cell (Sf21) expression system
Insect
GST