Learn More
Abnova™ Human DYRK4 (NP_003836.1, 1 a.a. - 520 a.a.) Full-length Recombinant Protein with Flag tag expressed in baculovirus infected Sf21 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P5543
Catalog No. 89-942-645
Add to cart
Description
Description
Dual-specificity kinases, such as DYRK4, play key roles in cell proliferation, survival, and development (Zhang et al., 2005 [PubMed 15607427]).[supplied by OMIM]
Specifications
Specifications
NP_003836.1
In 50mM Tris-HCl, 150mM NaCl, pH 7.5 (0.1% CHAPS, 1mM DTT, 10% glycerol)
61kDa
Baculovirus infected insect cell (Sf21) expression system
Loading 1 ug protein in SDS-PAGE
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DYRK4
Baculovirus infected insect cell (Sf21) expression system
Insect
Flag
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
8798
DYRK4 (Human) Recombinant Protein
60% by SDS-PAGE/CBB staining
5 ug
RUO
DYRK4
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg(or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip 3000. Substrate: DYRKtide. ATP: 100 uM.
Yes