Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human DYRK4 Partial ORF (AAH31244, 421 a.a. - 520 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00008798Q01L

 View more versions of this product

Catalog No. 89-942-644


Add to cart

Description

Description

Dual-specificity kinases, such as DYRK4, play key roles in cell proliferation, survival, and development (Zhang et al., 2005 [PubMed 15607427]).[supplied by OMIM]

Sequence: FFPSETRKDKVQGCHHSSRKADEITKETTEKTKDSPTKHVQHSGDQQDCLQHGADTVQLPQLVDAPKKSEAAVGAEVSMTSPGQSKNFSLKNTNVLPPIV
Specifications

Specifications

AAH31244
Liquid
8798
DYRK4 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DYRK4
wheat germ expression system
Yes
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
FFPSETRKDKVQGCHHSSRKADEITKETTEKTKDSPTKHVQHSGDQQDCLQHGADTVQLPQLVDAPKKSEAAVGAEVSMTSPGQSKNFSLKNTNVLPPIV
RUO
DYRK4
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit