Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human E4F1 Partial ORF (NP_004415.2, 685 a.a. - 784 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00001877Q01L

 View more versions of this product

Catalog No. 89-942-684


Add to cart

Description

Description

The zinc finger protein encoded by this gene is one of several cellular transcription factors whose DNA-binding activities are regulated through the action of adenovirus E1A. A 50-kDa amino-terminal product is generated from the full-length protein through proteolytic cleavage. The protein is differentially regulated by E1A-induced phosphorylation. The full-length gene product represses transcription from the E4 promoter in the absence of E1A, while the 50-kDa form acts as a transcriptional activator in its presence. [provided by RefSeq]

Sequence: GHQIIVQNVTMDEETALGPEAAAADTITIATPESLTEQVAMTLASAISEGTVLAARAGTSGTEQATVTMVSSEDIEILEHAGELVIASPEGQLEVQTVIV
Specifications

Specifications

NP_004415.2
Liquid
1877
E4F1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
E4F|MGC99614
E4F1
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GHQIIVQNVTMDEETALGPEAAAADTITIATPESLTEQVAMTLASAISEGTVLAARAGTSGTEQATVTMVSSEDIEILEHAGELVIASPEGQLEVQTVIV
RUO
E4F1
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit