Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human EBF4 Full-length ORF (AAH54347.1, 1 a.a. - 88 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | AAH54347.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 57593 |
Molecular Weight (g/mol) | 35.5kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-960-452
|
Abnova™
H00057593P01L |
25 ug |
Each for $769.00
|
|
|||||
89-960-451
|
Abnova™
H00057593P01S |
10 ug |
Each for $499.00
|
|
|||||
Description
EBF4 belongs to the conserved Olf/EBF family of helix-loop-helix transcription factors, members of which play important roles in neural development and B-cell maturation (Wang et al., 2002 [PubMed 12139918]).[supplied by OMIM]
Sequence: MPSSPPLAAASSMSLPAAAPTTSVFSFSPVNMISAVKQRSAFAPVLRPPSSPPQACPRAHGEGLPDQSFEDSDKFHSPARGLQGLAYSSpecifications
AAH54347.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
MPSSPPLAAASSMSLPAAAPTTSVFSFSPVNMISAVKQRSAFAPVLRPPSSPPQACPRAHGEGLPDQSFEDSDKFHSPARGLQGLAYS | |
RUO | |
EBF4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
57593 | |
EBF4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
COE4/KIAA1442/O/E-4 | |
EBF4 | |
Recombinant | |
wheat germ expression system |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title