Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ECEL1 Full-length ORF (AAH50453.2, 1 a.a. - 775 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00009427P01

Catalog No. 89-942-713


Add to cart

Description

Description

This gene encodes a member of the M13 family of endopeptidases. In general, M13 family members are zinc-containing type II integral-membrane proteins that are important regulators of neuropeptide and peptide hormone activity. This gene is expressed specifically in the central nervous system and its protein localizes predominately to the endoplasmic reticulum or, in trace amounts, to the cell surface. Disruption of this gene in mouse embryonic stem cells results in neonatal lethality due to respiratory failure shortly after birth. Based on the specific expression of this gene and the phenotype of the gene deficiency in mouse embryos, it is suggested that this protein plays a critical role in neural regulation of the respiratory system. This gene has multiple pseudogenes. [provided by RefSeq]

Sequence: MEPPYSLTAHYDEFQEVKYVSRCGAGGARGASLPPGFPLGAARSATGARSGLPRWNRREVCLLSGLVFAAGLCAILAAMLALKYLGPVAAGGGACPEGCPERKAFARAARFLAANLDASIDPCQDFYSFACGGWLRRHAIPDDKLTYGTIAAIGEQNEERLRRLLARPGGGPGGAAQRKVRAFFRSCLDMREIERLGPRPMLEVIEDCGGWDLGGAEERPGVAARWDLNRLLYKAQGVYSAAALFSLTVSLDDRNSSRYVIRIDQDGLTLPERTLYLAQDEDSEKILAAYRVFMERVLSLLGADAVEQKAQEILQVEQQLANITVSEYDDLRRDVSSMYNKVTLGQLQKITPHLRWKWLLDQIFQEDFSEEEEVVLLATDYMQQVSQLIRSTPHRVLHNYLVWRVVVVLSEHLSPPFREALHELAQEMEGSDKPQELARVCLGQANRHFGMALGALFVHEHFSAASKAKVQQLVEDIKYILGQRLEELDWMDAETRAAARAKLQYMMVMVGYPDFLLKPDAVDKEYEFEVHEKTYFKNILNSIRFSIQLSVKKIRQEVDKSTWLLPPQALNAYYLPNKNQMVFPAGILQPTLYDPDFPQSLNYGGIGTIIGHELTHGYDDWGGQYDRSGNLLHWWTEASYSRFLRKAECIVRLYDNFTVYNQRVNGKHTLGENIADMGGLKLAYHAYQKWVREHGPEHPLPRLKYTHDQLFFIAFAQNWCIKRRSQSIYLQVLTDKHAPEHYRVLGSVSQFEEFGRAFHCPKDSPMNPAHKCSVW
Specifications

Specifications

AAH50453.2
Liquid
9427
ECEL1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEPPYSLTAHYDEFQEVKYVSRCGAGGARGASLPPGFPLGAARSATGARSGLPRWNRREVCLLSGLVFAAGLCAILAAMLALKYLGPVAAGGGACPEGCPERKAFARAARFLAANLDASIDPCQDFYSFACGGWLRRHAIPDDKLTYGTIAAIGEQNEERLRRLLARPGGGPGGAAQRKVRAFFRSCLDMREIERLGPRPMLEVIEDCGGWDLGGAEERPGVAARWDLNRLLYKAQGVYSAAALFSLTVSLDDRNSSRYVIRIDQDGLTLPERTLYLAQDEDSEKILAAYRVFMERVLSLLGADAVEQKAQEILQVEQQLANITVSEYDDLRRDVSSMYNKVTLGQLQKITPHLRWKWLLDQIFQEDFSEEEEVVLLATDYMQQVSQLIRSTPHRVLHNYLVWRVVVVLSEHLSPPFREALHELAQEMEGSDKPQELARVCLGQANRHFGMALGALFVHEHFSAASKAKVQQLVEDIKYILGQRLEELDWMDAETRAAARAKLQYMMVMVGYPDFLLKPDAVDKEYEFEVHEKTYFKNILNSIRFSIQLSVKKIRQEVDKSTWLLPPQALNAYYLPNKNQMVFPAGILQPTLYDPDFPQSLNYGGIGTIIGHELTHGYDDWGGQYDRSGNLLHWWTEASYSRFLRKAECIVRLYDNFTVYNQRVNGKHTLGENIADMGGLKLAYHAYQKWVREHGPEHPLPRLKYTHDQLFFIAFAQNWCIKRRSQSIYLQVLTDKHAPEHYRVLGSVSQFEEFGRAFHCPKDSPMNPAHKCSVW
RUO
ECEL1
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
114.2kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DINE|ECEX|XCE
ECEL1
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit