Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ECH1 Full-length ORF (NP_001389.2, 1 a.a. - 328 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00001891P01S

 View more versions of this product

Catalog No. 89-942-719


Add to cart

Description

Description

This gene encodes a member of the hydratase/isomerase superfamily. The gene product shows high sequence similarity to enoyl-coenzyme A (CoA) hydratases of several species, particularly within a conserved domain characteristic of these proteins. The encoded protein, which contains a C-terminal peroxisomal targeting sequence, localizes to the peroxisome. The rat ortholog, which localizes to the matrix of both the peroxisome and mitochondria, can isomerize 3-trans,5-cis-dienoyl-CoA to 2-trans,4-trans-dienoyl-CoA, indicating that it is a delta3,5-delta2,4-dienoyl-CoA isomerase. This enzyme functions in the auxiliary step of the fatty acid beta-oxidation pathway. Expression of the rat gene is induced by peroxisome proliferators. [provided by RefSeq]

Sequence: MAAGIVASRRLRDLLTRRLTGSNYPGLSISLRLTGSSAQEEASGVALGEAPDHSYESLRVTSAQKHVLHVQLNRPNKRNAMNKVFWREMVECFNKISRDADCRAVVISGAGKMFTAGIDLMDMASDILQPKGDDVARISWYLRDIITRYQETFNVIERCPKPVIAAVHGGCIGGGVDLVTACDIRYCAQDAFFQVKEVDVGLAADVGTLQRLPKVIGNQSLVNELAFTARKMMADEALGSGLVSRVFPDKEVMLDAALALAAEISSKSPVAVQSTKVNLLYSRDHSVAESLNYVASWNMSMLQTQDLVKSVQATTENKELKTVTFSKL
Specifications

Specifications

NP_001389.2
Liquid
1891
ECH1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAAGIVASRRLRDLLTRRLTGSNYPGLSISLRLTGSSAQEEASGVALGEAPDHSYESLRVTSAQKHVLHVQLNRPNKRNAMNKVFWREMVECFNKISRDADCRAVVISGAGKMFTAGIDLMDMASDILQPKGDDVARISWYLRDIITRYQETFNVIERCPKPVIAAVHGGCIGGGVDLVTACDIRYCAQDAFFQVKEVDVGLAADVGTLQRLPKVIGNQSLVNELAFTARKMMADEALGSGLVSRVFPDKEVMLDAALALAAEISSKSPVAVQSTKVNLLYSRDHSVAESLNYVASWNMSMLQTQDLVKSVQATTENKELKTVTFSKL
RUO
ECH1
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
62.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HPXEL
ECH1
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit