Learn More
Abnova™ Human ECT2 Partial ORF (AAH06838.1, 46 a.a. - 145 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00001894Q01S
Description
The protein encoded by this gene is a transforming protein that is related to Rho-specific exchange factors and yeast cell cycle regulators. The expression of this gene is elevated with the onset of DNA synthesis and remains elevated during G2 and M phases. In situ hybridization analysis showed that expression is at a high level in cells undergoing mitosis in regenerating liver. Thus, this protein is expressed in a cell cycle-dependent manner during liver regeneration, and is thought to have an important role in the regulation of cytokinesis. [provided by RefSeq]
Sequence: LPKENWLKMLCRHVANTICKADAENLIYTADPESFEVNTKDMDSTLSRASRAIKKTSKKVTRAFSFSKTPKRALRRALMTSHGSVEGRSPSSNDKHVMSRSpecifications
AAH06838.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
LPKENWLKMLCRHVANTICKADAENLIYTADPESFEVNTKDMDSTLSRASRAIKKTSKKVTRAFSFSKTPKRALRRALMTSHGSVEGRSPSSNDKHVMSR | |
RUO | |
ECT2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
1894 | |
ECT2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ10461/MGC138291 | |
ECT2 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.