Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human EDA-A2/Ectodysplasin A2 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $468.00
Specifications
Antigen | EDA-A2/Ectodysplasin A2 |
---|---|
Concentration | LYOPH |
Dilution | Blockade of Receptor-ligand Interaction |
Applications | Blockade of Receptor-ligand Interaction |
Classification | Polyclonal |
Description
EDA-A2/Ectodysplasin A2 Polyclonal specifically detects EDA-A2/Ectodysplasin A2 in Human samples. It is validated for Blockade of Receptor-ligand Interaction.Specifications
EDA-A2/Ectodysplasin A2 | |
Blockade of Receptor-ligand Interaction | |
Polyclonal | |
Goat | |
Human | |
1896 | |
Mouse myeloma cell line NS0-derived recombinant human EDA-A2/Ectodysplasin A2 Ala179-Ser389 Accession # NP_001005609 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human EDA-A2/Ectodysplasin A2 in direct ELISAs and Western blots. In Western blots, less than 10% cross-reactivity with recombinant human (rh) APRIL, rhBAFF, rhLIGHT, rhOX40 Ligand, and rhTRAIL is observed and less than 2% cross-reactivity with recombinant mouse EDA, rhTNF-alpha , rhVEGI, rhGITR Ligand, and rhFasLigand is observed. |
LYOPH | |
Blockade of Receptor-ligand Interaction | |
Unconjugated | |
RUO | |
NP_001005609 | |
EDA | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title