Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EFHD1 Partial ORF (NP_079478.1, 168 a.a. - 238 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00080303Q01L

 View more versions of this product

Catalog No. 89-942-855


Add to cart

Description

Description

EFHD1 is an EF-hand domain-containing protein that displays increased expression during neuronal differentiation (Tominaga and Tomooka, 2002 [PubMed 12270117]).[supplied by OMIM]

Sequence: GLMALAKLSEIDVALEGVKGAKNFFEAKVQALSSASKFEAELKAEQDERKREEEERRLRQAAFQKLKANFN
Specifications

Specifications

NP_079478.1
Liquid
80303
EFHD1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp781H0842|FLJ13612|MST133|MSTP133|PP3051
EFHD1
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.44kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GLMALAKLSEIDVALEGVKGAKNFFEAKVQALSSASKFEAELKAEQDERKREEEERRLRQAAFQKLKANFN
RUO
EFHD1
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit