Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EFNA2 Full-length ORF (AAI48728.1, 1 a.a. - 213 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00001943P01L

 View more versions of this product

Catalog No. 89-942-868


Add to cart

Description

Description

This gene encodes a member of the ephrin family. The protein is composed of a signal sequence, a receptor-binding region, a spacer region, and a hydrophobic region. The EPH and EPH-related receptors comprise the largest subfamily of receptor protein-tyrosine kinases and have been implicated in mediating developmental events, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. Posttranslational modifications determine whether this protein localizes to the nucleus or the cytoplasm. [provided by RefSeq]

Sequence: MAPAQRPLLPLLLLLLPLPPPPFARAEDAARANSDRYAVYWNRSNPRFHAGAGDDGGGYTVEVSINDYLDIYCPHYGAPLPPAERMEHYVLYMVNGEGHASCDHRQRGFKRWECNRPAAPGGPLKFSEKFQLFTPFSLGFEFRPGHEYYYISATPPNAVDRPCLRLKVYVRPTNETLYEAPEPIFTSNNSCSSPGGCRLFLSTIPVLWTLLGS
Specifications

Specifications

AAI48728.1
Liquid
1943
EFNA2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAPAQRPLLPLLLLLLPLPPPPFARAEDAARANSDRYAVYWNRSNPRFHAGAGDDGGGYTVEVSINDYLDIYCPHYGAPLPPAERMEHYVLYMVNGEGHASCDHRQRGFKRWECNRPAAPGGPLKFSEKFQLFTPFSLGFEFRPGHEYYYISATPPNAVDRPCLRLKVYVRPTNETLYEAPEPIFTSNNSCSSPGGCRLFLSTIPVLWTLLGS
RUO
EFNA2
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
50.38kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ELF-1|EPLG6|HEK7-L|LERK6
EFNA2
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit