Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EFNB2 Full-length ORF (NP_004084.1, 1 a.a. - 333 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00001948P01S

 View more versions of this product

Catalog No. 89-942-882


Add to cart

Description

Description

This gene encodes a member of the ephrin (EPH) family. The ephrins and EPH-related receptors comprise the largest subfamily of receptor protein-tyrosine kinases and have been implicated in mediating developmental events, especially in the nervous system and in erythropoiesis. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. This gene encodes an EFNB class ephrin which binds to the EPHB4 and EPHA3 receptors. [provided by RefSeq]

Sequence: MAVRRDSVWKYCWGVLMVLCRTAISKSIVLEPIYWNSSNSKFLPGQGLVLYPQIGDKLDIICPKVDSKTVGQYEYYKVYMVDKDQADRCTIKKENTPLLNCAKPDQDIKFTIKFQEFSPNLWGLEFQKNKDYYIISTSNGSLEGLDNQEGGVCQTRAMKILMKVGQDASSAGSTRNKDPTRRPELEAGTNGRSSTTSPFVKPNPGSSTDGNSAGHSGNNILGSEVALFAGIASGCIIFIVIIITLVVLLLKYRRRHRKHSPQHTTTLSLSTLATPKRSGNNNGSEPSDIIIPLRTADSVFCPHYEKVSGDYGHPVYIVQEMPPQSPANIYYKV
Specifications

Specifications

NP_004084.1
Liquid
1948
EFNB2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAVRRDSVWKYCWGVLMVLCRTAISKSIVLEPIYWNSSNSKFLPGQGLVLYPQIGDKLDIICPKVDSKTVGQYEYYKVYMVDKDQADRCTIKKENTPLLNCAKPDQDIKFTIKFQEFSPNLWGLEFQKNKDYYIISTSNGSLEGLDNQEGGVCQTRAMKILMKVGQDASSAGSTRNKDPTRRPELEAGTNGRSSTTSPFVKPNPGSSTDGNSAGHSGNNILGSEVALFAGIASGCIIFIVIIITLVVLLLKYRRRHRKHSPQHTTTLSLSTLATPKRSGNNNGSEPSDIIIPLRTADSVFCPHYEKVSGDYGHPVYIVQEMPPQSPANIYYKV
RUO
EFNB2
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
63.3kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EPLG5|HTKL|Htk-L|LERK5|MGC126226|MGC126227|MGC126228
EFNB2
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit