Learn More
FUJIFILM IRVINE SCIENTIFIC INC Human EG-VEGF, Recombinant Protein, Shenandoah Biotechnology, 1 mg vial
Supplier:  FUJIFILM IRVINE SCIENTIFIC INC 100071MG
Encompass_Preferred
Endocrine gland-derived vascular endothelial growth factor (EG-VEGF) is an angiogenic growth factor that is expressed in the ovaries, testis, adrenal, and placental tissues.   EG-VEGF has mitogenic, chemoattractive, and antiapoptotic functional roles.  EG-VEGF signaling is mediated through binding the G protein-coupled receptors prokineticin receptor 1 (PKR1) and prokineticin receptor 2 (PKR2).  Polycystic ovaries display strong EG-VEGF expression that is associated with increased angiogenesis and cyst formation, which could lead to the formation of polycystic ovary syndrome and infertility. 
Catalog No. 50-900-460
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.