Learn More
Bon Opus BiosciencesSupplier Diversity Partner Recombinant Human Epidermal Growth Factor/EGF  DFS Item
Manufacturer:  Bon Opus BiosciencesSupplier Diversity Partner C02910UG
Recombinant Human Epidermal Growth Factor is produced by our E.coli expression system and the target gene encoding Asn971-Arg1023 is expressed. Bon Opus Cat. #C029
Catalog No. 50-719-959