Learn More
Abnova™ Human EGF (P01133) Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4381
Catalog No. 89-942-896
Add to cart
Description
Description
Epidermal growth factor has a profound effect on the differentiation of specific cells in vivo and is a potent mitogenic factor for a variety of cultured cells of both ectodermal and mesodermal origin. The EGF precursor is believed to exist as a membrane-bound molecule which is proteolytically cleaved to generate the 53-amino acid peptide hormone that stimulates cells to divide. [provided by RefSeq]
Sequence: NSDSECPLSHDGYCLHDGVCMYIEALDKYACNCVVGYIGERCQYRDLKWWELR
Specifications
Specifications
P01133
Lyophilized
1950
EGF (Human) Recombinant Protein
500 ug
Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
<0.1 EU/μg
EGF
Escherichia coli expression system
Escherichia coli
None
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
6.2kDa
Escherichia coli expression system
NSDSECPLSHDGYCLHDGVCMYIEALDKYACNCVVGYIGERCQYRDLKWWELR
RUO
HOMG4|URG
EGF
The activity is determined by the dose-dependent proliferation of murine BALB/c 3T3 cells. The expected ED50 for this effect is 16-24pg/mL.
Yes