Learn More
Abnova™ Human EGF (Q6QBS2, 53 amino acids) Partial Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P3960
Catalog No. 89-942-895
Add to cart
Description
Description
Epidermal growth factor has a profound effect on the differentiation of specific cells in vivo and is a potent mitogenic factor for a variety of cultured cells of both ectodermal and mesodermal origin. The EGF precursor is believed to exist as a membrane-bound molecule which is proteolytically cleaved to generate the 53-amino acid peptide hormone that stimulates cells to divide. [provided by RefSeq]