Learn More
Abnova™ Human EGFR (NM_005228.3, 672 a.a. - 1210 a.a.) G719S mutant Partial Protein expressed in Sf9 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4814
Catalog No. 89-942-923
Add to cart
Description
Description
The protein encoded by this gene is a transmembrane glycoprotein that is a member of the protein kinase superfamily. This protein is a receptor for members of the epidermal growth factor family. EGFR is a cell surface protein that binds to epidermal growth factor. Binding of the protein to a ligand induces receptor dimerization and tyrosine autophosphorylation and leads to cell proliferation. Mutations in this gene are associated with lung cancer. [provided by RefSeq]
Specifications
Specifications
NM_005228.3
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
89.201kDa
Insect cell (Sf9) expression system
2 ug/lane SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing
ERBB|ERBB1|HER1|PIG61|mENA
EGFR
54pmol/μg x min
None
0.367 ug/uL
Liquid
1956
EGFR (G719S) (Human) Recombinant Protein
GST affinity chromatography
100 ug
RUO
EGFR
Insect cell (Sf9) expression system
Insect