Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EHF Full-length ORF (NP_036285.2, 1 a.a. - 300 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00026298P01S

 View more versions of this product

Catalog No. 89-942-966


Add to cart

Description

Description

This gene encodes a protein that belongs to an ETS transcription factor subfamily characterized by epithelial-specific expression (ESEs). The encoded protein acts as a transcriptional repressor and may be associated with asthma susceptibility. This protein may be involved in epithelial differentiation and carcinogenesis. [provided by RefSeq]

Sequence: MILEGGGVMNLNPGNNLLHQPPAWTDSYSTCNVSSGFFGGQWHEIHPQYWTKYQVWEWLQHLLDTNQLDANCIPFQEFDINGEHLCSMSLQEFTRAAGTAGQLLYSNLQHLKWNGQCSSDLFQSTHNVIVKTEQTEPSIMNTWKDENYLYDTNYGSTVDLLDSKTFCRAQISMTTTSHLPVAESPDMKKEQDPPAKCHTKKHNPRGTHLWEFIRDILLNPDKNPGLIKWEDRSEGVFRFLKSEAVAQLWGKKKNNSSMTYEKLSRAMRYYYKREILERVDGRRLVYKFGKNARGWRENEN
Specifications

Specifications

NP_036285.2
Liquid
26298
EHF (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MILEGGGVMNLNPGNNLLHQPPAWTDSYSTCNVSSGFFGGQWHEIHPQYWTKYQVWEWLQHLLDTNQLDANCIPFQEFDINGEHLCSMSLQEFTRAAGTAGQLLYSNLQHLKWNGQCSSDLFQSTHNVIVKTEQTEPSIMNTWKDENYLYDTNYGSTVDLLDSKTFCRAQISMTTTSHLPVAESPDMKKEQDPPAKCHTKKHNPRGTHLWEFIRDILLNPDKNPGLIKWEDRSEGVFRFLKSEAVAQLWGKKKNNSSMTYEKLSRAMRYYYKREILERVDGRRLVYKFGKNARGWRENEN
RUO
EHF
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
61.3kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ESE3|ESEJ
EHF
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit