Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human eIF2 alpha Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB3997
This item is not returnable.
View return policy
Description
eIF2 alpha/EIF2S1 Monoclonal specifically detects eIF2 alpha/EIF2S1 in Human samples. It is validated for Western Blot.Specifications
eIF2 alpha/EIF2S1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q53XC0 | |
EIF2S1 | |
E. coli-derived recombinant human eIF2 alpha Met1-Asp315 Accession # Q53XC0 | |
100 μg | |
Primary | |
Detects human eIF2 alpha in Western blots. | |
Human | |
Purified |
Western Blot | |
430516 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EIF2, EIF-2, EIF2A, EIF-2A, EIF-2alpha, EIF2S1, EIF2S1 eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1965 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction