Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EIF2AK1 (BAC11461.1, 1 a.a. - 630 a.a.) Full length Recombinant Protein with GST-tag

Used for Func, SDS-PAGE

Manufacturer:  Abnova™ P5769

Catalog No. 89-942-986


Add to cart

Description

Description

The protein encoded by this gene acts at the level of translation initiation to downregulate protein synthesis in response to stress. The encoded protein is a kinase that can be inactivated by hemin. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Specifications

BAC11461.1
Liquid
27102
EIF2AK1 (Human) Recombinant Protein
Glutathione sepharose chromatography
SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HCR|HRI|KIAA1369
EIF2AK1
The activity was determined by ELISA. The enzyme was incubated with GST-fused substrate protein, and after stopping kinase reaction by EDTA, the reaction solution was transferred into glutathione-coated plate. Phosphorylation was detected by anti-phospho antibody and HRP-labeled anti-rabbit IgG.
Substrate : EIF2S1. ATP: 100 µM.
Yes
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
98kDa
Escherichia coli expression system
71% by SDS-PAGE/CBB staining
5 ug
RUO
EIF2AK1
Escherichia coli expression system
Escherichia coli
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit