Learn More
Abnova™ Human EIF2AK1 (NM_014413.2, 1 a.a. - 630 a.a.) Full-length Recombinant Protein with GST-His tag expressed in Sf9 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4701
Catalog No. 89-942-985
Add to cart
Description
Description
The protein encoded by this gene acts at the level of translation initiation to downregulate protein synthesis in response to stress. The encoded protein is a kinase that can be inactivated by hemin. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Specifications
Specifications
NM_014413.2
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
100.546kDa
Insect cell (Sf9) expression system
2 ug/lane SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing
HCR|HRI|KIAA1369
EIF2AK1
3pmol/μg x min
Yes
0.030 ug/uL
Liquid
27102
EIF2AK1 (Human) Recombinant Protein
GST affinity chromatography
100 ug
RUO
EIF2AK1
Insect cell (Sf9) expression system
Insect
GST-His