Learn More
Abnova™ Human EIF2AK2 (NP_002750.1, 252 a.a. - 551 a.a.) Partial Recombinant Protein with GST-tag expressed in baculovirus infected Sf21 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P5626
Catalog No. 89-942-992
Add to cart
Specifications
Specifications
NP_002750.1
Liquid
5610
EIF2AK2 (Human) Recombinant Protein
Glutathione sepharose chromatography and anion exchange chromatography
Loading 1 ug protein in SDS-PAGE
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EIF2AK1|MGC126524|PKR|PRKR
EIF2AK2
The activity was determined by IMAPTM assay. The enzyme was incubated with fluorescein labeled peptide and phosphorylation was detected by IMAPTM technology (fluorescence polarization). Substrate: SRPKtide. ATP: 100μM.
Yes
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
62kDa
Baculovirus infected insect cell (Sf21) expression system
61% by SDS-PAGE/CBB staining
5 ug
RUO
EIF2AK2
Baculovirus infected insect cell (Sf21) expression system
Insect
GST