Learn More
Abnova™ Human EIF5B Partial ORF (NP_056988, 1121 a.a. - 1218 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_056988 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9669 |
Molecular Weight (g/mol) | 36.52kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-943-106
|
Abnova™
H00009669Q01S |
10 ug |
Each for $499.00
|
|
|||||
89-943-107
|
Abnova™
H00009669Q01L |
25 ug |
Each for $769.00
|
|
|||||
Description
Accurate initiation of translation in eukaryotes is complex and requires many factors, some of which are composed of multiple subunits. The process is simpler in prokaryotes which have only three initiation factors (IF1, IF2, IF3). Two of these factors are conserved in eukaryotes: the homolog of IF1 is eIF1A and the homolog of IF2 is eIF5B. This gene encodes eIF5B. Factors eIF1A and eIF5B interact on the ribosome along with other initiation factors and GTP to position the initiation methionine tRNA on the start codon of the mRNA so that translation initiates accurately. [provided by RefSeq]
Sequence: QGTPMCVPSKNFVDIGIVTSIEINHKQVDVAKKGQEVCVKIEPIPGESPKMFGRHFEATDILVSKISRQSIDALKDWFRDEMQKSDWQLIVELKKVFESpecifications
NP_056988 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434I036/FLJ10524/IF2/KIAA0741 | |
EIF5B | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9669 | |
EIF5B (Human) Recombinant Protein (Q01) | |
QGTPMCVPSKNFVDIGIVTSIEINHKQVDVAKKGQEVCVKIEPIPGESPKMFGRHFEATDILVSKISRQSIDALKDWFRDEMQKSDWQLIVELKKVFE | |
RUO | |
EIF5B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.