Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ELA1 Partial ORF (NP_001962.3, 159 a.a. - 257 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00001990Q01S

 View more versions of this product

Catalog No. 89-943-110


Add to cart

Description

Description

Elastases form a subfamily of serine proteases that hydrolyze many proteins in addition to elastin. Humans have six elastase genes which encode the structurally similar proteins elastase 1, 2, 2A, 2B, 3A, and 3B. Unlike other elastases, pancreatic elastase 1 is not expressed in the pancreas. To date, elastase 1 expression has only been detected in skin keratinocytes. Clinical literature that describes human elastase 1 activity in the pancreas or fecal material is actually referring to chymotrypsin-like elastase family, member 3B. [provided by RefSeq]

Sequence: LAQTLQQAYLPSVDYAICSSSSYWGSTVKNTMVCAGGDGVRSGCQGDSGGPLHCLVNGKYSVHGVTSFVSSRGCNVSRKPTVFTQVSAYISWINNVIAS
Specifications

Specifications

NP_001962.3
Liquid
1990
ELA1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ELA1
wheat germ expression system
Yes
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LAQTLQQAYLPSVDYAICSSSSYWGSTVKNTMVCAGGDGVRSGCQGDSGGPLHCLVNGKYSVHGVTSFVSSRGCNVSRKPTVFTQVSAYISWINNVIAS
RUO
ELA1
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit