Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ELAC2 Full-length ORF (NP_060597.3, 1 a.a. - 826 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00060528P01

Catalog No. 89-943-128


Add to cart

Description

Description

The protein encoded by this gene has a C-terminal domain with tRNA 3′ processing endoribonuclease activity, which catalyzes the removal of the 3' trailer from precursor tRNAs. The protein also interacts with activated Smad family member 2 (Smad2) and its nuclear partner forkhead box H1 (also known as FAST-1), and reduced expression can suppress transforming growth factor-beta induced growth arrest. Mutations in this gene result in an increased risk of prostate cancer. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MWALCSLLRSAAGRTMSQGRTISQAPARRERPRKDPLRHLRTREKRGPSGCSGGPNTVYLQVVAAGSRDSGAALYVFSEFNRYLFNCGEGVQRLMQEHKLKVARLDNIFLTRMHWSNVGGLSGMILTLKETGLPKCVLSGPPQLEKYLEAIKIFSGPLKGIELAVRPHSAPEYEDETMTVYQIPIHSEQRRGKHQPWQSPERPLSRLSPERSSDSELNENEPHLPHGVSQRRGVRDSSLVVAFICKLHLKRGNFLVLKAKEMGLPVGTAAIAPIIAAVKDGKSITHEGREILAEELCTPPDPGAAFVVVECPDESFIQPICENATFQRYQGKADAPVALVVHMAPASVLVDSRYQQWMERFGPDTQHLVLNENCASVHNLRSHKIQTQLNLIHPDIFPLLTSFRCKKEGPTLSVPMVQGECLLKYQLRPRREWQRDAIITCNPEEFIVEALQLPNFQQSVQEYRRSAQDGPAPAEKRSQYPEIIFLGTGSAIPMKIRNVSATLVNISPDTSLLLDCGEGTFGQLCRHYGDQVDRVLGTLAAVFVSHLHADHHTGLPSILLQRERALASLGKPLHPLLVVAPNQLKAWLQQYHNQCQEVLHHISMIPAKCLQEGAEISSPAVERLISSLLRTCDLEEFQTCLVRHCKHAFGCALVHTSGWKVVYSGDTMPCEALVRMGKDATLLIHEATLEDGLEEEAVEKTHSTTSQAISVGMRMNAEFIMLNHFSQRYAKVPLFSPNFSEKVGVAFDHMKVCFGDFPTMPKLIPPLKALFAGDIEEMEERREKRELRQVRAALLSRELAGGLEDGEPQQKRAHTEEPQAKKVRAQ
Specifications

Specifications

NP_060597.3
Liquid
60528
ELAC2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MWALCSLLRSAAGRTMSQGRTISQAPARRERPRKDPLRHLRTREKRGPSGCSGGPNTVYLQVVAAGSRDSGAALYVFSEFNRYLFNCGEGVQRLMQEHKLKVARLDNIFLTRMHWSNVGGLSGMILTLKETGLPKCVLSGPPQLEKYLEAIKIFSGPLKGIELAVRPHSAPEYEDETMTVYQIPIHSEQRRGKHQPWQSPERPLSRLSPERSSDSELNENEPHLPHGVSQRRGVRDSSLVVAFICKLHLKRGNFLVLKAKEMGLPVGTAAIAPIIAAVKDGKSITHEGREILAEELCTPPDPGAAFVVVECPDESFIQPICENATFQRYQGKADAPVALVVHMAPASVLVDSRYQQWMERFGPDTQHLVLNENCASVHNLRSHKIQTQLNLIHPDIFPLLTSFRCKKEGPTLSVPMVQGECLLKYQLRPRREWQRDAIITCNPEEFIVEALQLPNFQQSVQEYRRSAQDGPAPAEKRSQYPEIIFLGTGSAIPMKIRNVSATLVNISPDTSLLLDCGEGTFGQLCRHYGDQVDRVLGTLAAVFVSHLHADHHTGLPSILLQRERALASLGKPLHPLLVVAPNQLKAWLQQYHNQCQEVLHHISMIPAKCLQEGAEISSPAVERLISSLLRTCDLEEFQTCLVRHCKHAFGCALVHTSGWKVVYSGDTMPCEALVRMGKDATLLIHEATLEDGLEEEAVEKTHSTTSQAISVGMRMNAEFIMLNHFSQRYAKVPLFSPNFSEKVGVAFDHMKVCFGDFPTMPKLIPPLKALFAGDIEEMEERREKRELRQVRAALLSRELAGGLEDGEPQQKRAHTEEPQAKKVRAQ
RUO
ELAC2
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
118.6kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ELC2|FLJ10530|FLJ36693|FLJ42848|HPC2
ELAC2
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit