Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ELK3 Full-length ORF (NP_005221.2, 1 a.a. - 407 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00002004P01

Catalog No. 89-943-169


Add to cart

Description

Description

The protein encoded by this gene is a member of the ETS-domain transcription factor family and the ternary complex factor (TCF) subfamily. Proteins in this subfamily regulate transcription when recruited by serum response factor to bind to serum response elements. This protein is activated by signal-induced phosphorylation; studies in rodents suggest that it is a transcriptional inhibitor in the absence of Ras, but activates transcription when Ras is present. [provided by RefSeq]

Sequence: MESAITLWQFLLQLLLDQKHEHLICWTSNDGEFKLLKAEEVAKLWGLRKNKTNMNYDKLSRALRYYYDKNIIKKVIGQKFVYKFVSFPEILKMDPHAVEISRESLLLQDSDCKASPEGREAHKHGLAALRSTSRNEYIHSGLYSSFTINSLQNPPDAFKAIKTEKLEEPPEDSPPVEEVRTVIRFVTNKTDKHVTRPVVSLPSTSEAAAASAFLASSVSAKISSLMLPNAASISSASPFSSRSPSLSPNSPLPSEHRSLFLEAACHDSDSLEPLNLSSGSKTKSPSLPPKAKKPKGLEISAPPLVLSGTDIGSIALNSPALPSGSLTPAFFTAQTPNGLLLTPSPLLSSIHFWSSLSPVAPLSPARLQGPSTLFQFPTLLNGHMPVPIPSLDRAASPVLLSSNSQKS
Specifications

Specifications

NP_005221.2
Liquid
2004
ELK3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MESAITLWQFLLQLLLDQKHEHLICWTSNDGEFKLLKAEEVAKLWGLRKNKTNMNYDKLSRALRYYYDKNIIKKVIGQKFVYKFVSFPEILKMDPHAVEISRESLLLQDSDCKASPEGREAHKHGLAALRSTSRNEYIHSGLYSSFTINSLQNPPDAFKAIKTEKLEEPPEDSPPVEEVRTVIRFVTNKTDKHVTRPVVSLPSTSEAAAASAFLASSVSAKISSLMLPNAASISSASPFSSRSPSLSPNSPLPSEHRSLFLEAACHDSDSLEPLNLSSGSKTKSPSLPPKAKKPKGLEISAPPLVLSGTDIGSIALNSPALPSGSLTPAFFTAQTPNGLLLTPSPLLSSIHFWSSLSPVAPLSPARLQGPSTLFQFPTLLNGHMPVPIPSLDRAASPVLLSSNSQKS
RUO
ELK3
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
70.6kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ERP|NET|SAP2
ELK3
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit