Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human ELMO2 Partial ORF (NP_573403.1, 89 a.a. - 188 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00063916Q01S
Description
The protein encoded by this gene interacts with the dedicator of cyto-kinesis 1 protein. Similarity to a C. elegans protein suggests that this protein may function in phagocytosis of apoptotic cells and in cell migration. Alternative splicing results in multiple transcript variants encoding the same protein. [provided by RefSeq]
Sequence: MERTQSSNMETRLDAMKELAKLSADVTFATEFINMDGIIVLTRLVESGTKLLSHYSEMLAFTLTAFLELMDHGIVSWDMVSITFIKQIAGYVSQPMVDVSSpecifications
NP_573403.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MERTQSSNMETRLDAMKELAKLSADVTFATEFINMDGIIVLTRLVESGTKLLSHYSEMLAFTLTAFLELMDHGIVSWDMVSITFIKQIAGYVSQPMVDVS | |
RUO | |
ELMO2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
63916 | |
ELMO2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CED-12/CED12/ELMO-2/FLJ11656/KIAA1834 | |
ELMO2 | |
Recombinant | |
wheat germ expression system |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction