Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ELMO3 Partial ORF (AAH34410.1, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00079767Q01S

 View more versions of this product

Catalog No. 89-943-184


Add to cart

Description

Description

The protein encoded by this gene is similar to a C. elegans protein that functions in phagocytosis of apoptotic cells and in cell migration. Other members of this small family of engulfment and cell motility (ELMO) proteins have been shown to interact with the dedicator of cyto-kinesis 1 protein to promote phagocytosis and effect cell shape changes. [provided by RefSeq]

Sequence: MIFAREVISRNGLQILGTIIEDGDDLGEVLALSLRAFSELMEHGVVSWETLSIPFVRKVVCYVNMNLMDASVPPLALGLLESVTLSSPALGQLVKSEVPL
Specifications

Specifications

AAH34410.1
Liquid
79767
ELMO3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CED-12|CED12|ELMO-3|FLJ13824
ELMO3
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MIFAREVISRNGLQILGTIIEDGDDLGEVLALSLRAFSELMEHGVVSWETLSIPFVRKVVCYVNMNLMDASVPPLALGLLESVTLSSPALGQLVKSEVPL
RUO
ELMO3
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit