Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EMCN (NP_057326, 19 a.a. - 190 a.a.) Partial Recombinant Protein with His tag

Used for SDS-PAGE

Manufacturer:  Abnova™ P5853

Catalog No. 89-943-212


Add to cart

Description

Description

EMCN is a mucin-like sialoglycoprotein that interferes with the assembly of focal adhesion complexes and inhibits interaction between cells and the extracellular matrix (Kinoshita et al., 2001 [PubMed 11418125]).[supplied by OMIM]

Sequence: MGSSHHHHHHSSGLVPRGSHMGSHMNSTGVLEAANNSLVVTTTKPSITTPNTESLQKNVVTPTTGTTPKGTITNELLKMSLMSTATFLTSKDEGLKATTTDVRKNDSIISNVTVTSVTLPNAVSTLQSSKPKTETQSSIKTTEIPGSVLQPDASPSKTGTLTSIPVTIPENTSQSQVIGTEGGKNASTSATSRSYSS
Specifications

Specifications

Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
20.5kDa
Escherichia coli expression system
3 ug/lane in 15% SDS-PAGE Stained with Coomassie Blue. Due to the protein nature, dimmers and multimers may be observed.
MGSSHHHHHHSSGLVPRGSHMGSHMNSTGVLEAANNSLVVTTTKPSITTPNTESLQKNVVTPTTGTTPKGTITNELLKMSLMSTATFLTSKDEGLKATTTDVRKNDSIISNVTVTSVTLPNAVSTLQSSKPKTETQSSIKTTEIPGSVLQPDASPSKTGTLTSIPVTIPENTSQSQVIGTEGGKNASTSATSRSYSS
RUO
EMCN
Escherichia coli expression system
Yes
Liquid
51705
EMCN (Human) Recombinant Protein
>90% by SDS - PAGE
20 ug
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
EMCN2|MUC14
EMCN
Escherichia coli
His
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit