Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EME1 Full-length ORF (AAH16470.1, 1 a.a. - 583 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00146956P01L

 View more versions of this product

Catalog No. 89-943-218


Add to cart

Description

Description

This gene encodes a protein that complexes with methyl methanesulfonate-sensitive UV-sensitive 81 protein to form an endonuclease complex. The encoded protein interacts with specifc DNA structures including nicked Holliday junctions, 3'-flap structures and aberrant replication fork structures. This protein may be involved in repairing DNA damage and in maintaining genomic stability

Sequence: MALKKSSPSLDSGDSDSEELPTFAFLKKEPSSTKRRQPEREEKIVVVDISDCEASCPPAPELFSPPVPDIAETVTQTQPVRLLSSESEDEEEFIPLAQRLTCKFLTHKQLSPEDSSSPVKSVLDHQNNEGASCDWKKPFPKIPEVPLHDTPERSAADNKDLILDPCCQLPAYLSTCPGQSSSLAVTKTNSDILPPQKKTKPSQKVQGRGSHGCRQQRQARQKESTLRRQERKNAALVTRMKAQRPEECLKHIIVVLDPVLLQMEGGGQLLGALQTMECRCVIEAQAVPCSVTWRRRAGPSEDREDWVEEPTVLVLLRAEAFVSMIDNGKQGSLDSTMKGKETLQGFVTDITAKTAGKALSLVIVDQEKCFSLELLFFDFLPCTSAQNPPRRGKQGANKQTKKQQQRQPEASIGSMVSRVDAEEALVDLQLHTEAQAQIVQSWKELADFTCAFTKAVAEAPFKKLRDETTFSFCLESDWAGGVKVDLAGRGLALVWRRQIQQLNRVSLEMASAVVNAYPSPQLLVQAYQQCFSDKERQNLLADIQVRRGEGVTSTSRRIGPELSRRIYLQMTTLQPHLSLDSAD
Specifications

Specifications

AAH16470.1
Liquid
146956
EME1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MALKKSSPSLDSGDSDSEELPTFAFLKKEPSSTKRRQPEREEKIVVVDISDCEASCPPAPELFSPPVPDIAETVTQTQPVRLLSSESEDEEEFIPLAQRLTCKFLTHKQLSPEDSSSPVKSVLDHQNNEGASCDWKKPFPKIPEVPLHDTPERSAADNKDLILDPCCQLPAYLSTCPGQSSSLAVTKTNSDILPPQKKTKPSQKVQGRGSHGCRQQRQARQKESTLRRQERKNAALVTRMKAQRPEECLKHIIVVLDPVLLQMEGGGQLLGALQTMECRCVIEAQAVPCSVTWRRRAGPSEDREDWVEEPTVLVLLRAEAFVSMIDNGKQGSLDSTMKGKETLQGFVTDITAKTAGKALSLVIVDQEKCFSLELLFFDFLPCTSAQNPPRRGKQGANKQTKKQQQRQPEASIGSMVSRVDAEEALVDLQLHTEAQAQIVQSWKELADFTCAFTKAVAEAPFKKLRDETTFSFCLESDWAGGVKVDLAGRGLALVWRRQIQQLNRVSLEMASAVVNAYPSPQLLVQAYQQCFSDKERQNLLADIQVRRGEGVTSTSRRIGPELSRRIYLQMTTLQPHLSLDSAD
RUO
EME1
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
91.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ31364|MMS4L
EME1
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit