Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PKN3 Full-length ORF (NP_037487.2, 1 a.a. - 889 a.a.) Recombinant Protein with GST-tag at N-terminal (P02)

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00002009P01

Catalog No. 89-943-230


Add to cart

Description

Description

Human echinoderm microtubule-associated protein-like is a strong candidate for the Usher syndrome type 1A gene. Usher syndromes (USHs) are a group of genetic disorders consisting of congenital deafness, retinitis pigmentosa, and vestibular dysfunction of variable onset and severity depending on the genetic type. The disease process in USHs involves the entire brain and is not limited to the posterior fossa or auditory and visual systems. The USHs are catagorized as type I (USH1A, USH1B, USH1C, USH1D, USH1E and USH1F), type II (USH2A and USH2B) and type III (USH3). The type I is the most severe form. Gene loci responsible for these three types are all mapped. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MEDGFSSYSSLYDTSSLLQFCNDDSASAASSMEVTDRIASLEQRVQMQEDDIQLLKSALADVVRRLNITEEQQAVLNRKGPTKARPLMQTLPLRTTVNNGTVLPKKPTGSLPSPSGVRKETAVPATKSNIKRTSSSERVSPGGRRESNGDSRGNRNRTGSTSSSSSGKKNSESKPKEPVFSAEEGYVKMFLRGRPVTMYMPKDQVDSYSLEAKVELPTKRLKLEWVYGYRGRDCRNNLYLLPTGETVYFIASVVVLYNVEEQLQRHYAGHNDDVKCLAVHPDRITIATGQVAGTSKDGKQLPPHVRIWDSVTLNTLHVIGIGFFDRAVTCIAFSKSNGGTNLCAVDDSNDHVLSVWDWQKEEKLADVKCSNEAVFAADFHPTDTNIIVTCGKSHLYFWTLEGSSLNKKQGLFEKQEKPKFVLCVTFSENGDTITGDSSGNILVWGKGTNRISYAVQGAHEGGIFALCMLRDGTLVSGGGKDRKLISWSGNYQKLRKTEIPEQFGPIRTVAEGKGDVILIGTTRNFVLQGTLSGDFTPITQGHTDELWGLAINASKSQFLTCGHDKHATLWDAVGHRPVWDKIIEDPAQSSGFHPSGSVVAVGTLTGRWFVFDTGTKDLVTVHTDGNEQLSVMRYSPDGNFLAIGSHDNCIYIYGVSDNGRKYTRVGKCSGHSSFITHLDWSVNSQFLVSNSGDYEILYWVPSACKQVVSVETTRDIEWATYTCTLGFHVFGVWPEGSDGTDINAVCRAHEKKLLSTGDDFGKVHLFSYPCSQFRAPSHIYGGHSSHVTNVDFLCEDSHLISTGGKDTSIMQWRVI
Specifications

Specifications

AAH33043.1
Liquid
2009
EML1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEDGFSSYSSLYDTSSLLQFCNDDSASAASSMEVTDRIASLEQRVQMQEDDIQLLKSALADVVRRLNITEEQQAVLNRKGPTKARPLMQTLPLRTTVNNGTVLPKKPTGSLPSPSGVRKETAVPATKSNIKRTSSSERVSPGGRRESNGDSRGNRNRTGSTSSSSSGKKNSESKPKEPVFSAEEGYVKMFLRGRPVTMYMPKDQVDSYSLEAKVELPTKRLKLEWVYGYRGRDCRNNLYLLPTGETVYFIASVVVLYNVEEQLQRHYAGHNDDVKCLAVHPDRITIATGQVAGTSKDGKQLPPHVRIWDSVTLNTLHVIGIGFFDRAVTCIAFSKSNGGTNLCAVDDSNDHVLSVWDWQKEEKLADVKCSNEAVFAADFHPTDTNIIVTCGKSHLYFWTLEGSSLNKKQGLFEKQEKPKFVLCVTFSENGDTITGDSSGNILVWGKGTNRISYAVQGAHEGGIFALCMLRDGTLVSGGGKDRKLISWSGNYQKLRKTEIPEQFGPIRTVAEGKGDVILIGTTRNFVLQGTLSGDFTPITQGHTDELWGLAINASKSQFLTCGHDKHATLWDAVGHRPVWDKIIEDPAQSSGFHPSGSVVAVGTLTGRWFVFDTGTKDLVTVHTDGNEQLSVMRYSPDGNFLAIGSHDNCIYIYGVSDNGRKYTRVGKCSGHSSFITHLDWSVNSQFLVSNSGDYEILYWVPSACKQVVSVETTRDIEWATYTCTLGFHVFGVWPEGSDGTDINAVCRAHEKKLLSTGDDFGKVHLFSYPCSQFRAPSHIYGGHSSHVTNVDFLCEDSHLISTGGKDTSIMQWRVI
RUO
EML1
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
115.39kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ELP79|EMAP|EMAPL|FLJ45033|HuEMAP
EML1
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit