Learn More
Abnova™ Human EMR4 Partial ORF (XP_377506, 21 a.a. - 93 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00326342Q01S
Description
This gene is a member of the EGF-TM7 receptor gene family which is thought to play a role in leukocyte adhesion and migration. In other vertebrates, including nonhuman primates, this gene encodes a protein containing N-terminal EGF domains and a C-terminal transmembrane domain. Sequence evidence for the human gene, however, indicates nucleotide deletion in the genomic sequence would result in frameshift and early termination of translation. A protein expressed by this gene would be soluble rather than expressed on the cell surface. As the encoded protein has not been detected, this gene may represent a transcribed pseudogene. [provided by RefSeq]
Sequence: GSEAKNSGASCPPCPKYASCHNSTHCTCEDGFRARSGRTYFHDSSEKCEDINECETGLAKCKYKAYCRNKVGGSpecifications
XP_377506 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.77kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GSEAKNSGASCPPCPKYASCHNSTHCTCEDGFRARSGRTYFHDSSEKCEDINECETGLAKCKYKAYCRNKVGG | |
RUO | |
EMR4P | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
326342 | |
EMR4 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EMR4/FIRE/GPR127/PGR16 | |
EMR4P | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.