Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ENAM Partial ORF (NP_114095, 1043 a.a. - 1141 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00010117Q01S

 View more versions of this product

Catalog No. 89-943-252


Add to cart

Description

Description

Dental enamel is a highly mineralized tissue with 85% of its volume occupied by unusually large, highly organized, hydroxyapatite crystals. This highly organized and unusual structure is thought to be rigorously controlled in ameloblasts through the interaction of a number of organic matrix molecules that include enamelin, amelogenin (AMELX; MIM 300391), ameloblastin (AMBN; MIM 601259), tuftelin (TUFT1; MIM 600087), dentine sialophosphoprotein (DSPP; MIM 125485), and a variety of enzymes. Enamelin is the largest protein in the enamel matrix of developing teeth and comprises approximately 5% of total enamel matrix protein.[supplied by OMIM]

Sequence: ERQQQRPSNILHLPCFGSKLAKHHSSTTGTPSSDGRQSPFDGDSITPTENPNTLVELATEEQFKSINVDPLDADEHSPFEFLQRGTNVQDQVQDCLLLQ
Specifications

Specifications

NP_114095
Liquid
10117
ENAM (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ADAI|AI1C|AIH2
ENAM
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ERQQQRPSNILHLPCFGSKLAKHHSSTTGTPSSDGRQSPFDGDSITPTENPNTLVELATEEQFKSINVDPLDADEHSPFEFLQRGTNVQDQVQDCLLLQ
RUO
ENAM
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit