Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ENC1 Full-length ORF (NP_003624.1, 1 a.a. - 589 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00008507P01S

 View more versions of this product

Catalog No. 89-943-256


Add to cart

Description

Description

DNA damage and/or hyperproliferative signals activate wildtype p53 tumor suppressor protein (TP53; MIM 191170), inducing cell cycle arrest or apoptosis. Mutations that inactivate p53 occur in 50% of all tumors. Polyak et al. (1997) [PubMed 9305847] used serial analysis of gene expression (SAGE) to evaluate cellular mRNA levels in a colorectal cancer cell line transfected with p53. Of 7,202 transcripts identified, only 14 were expressed at levels more than 10-fold higher in p53-expressing cells than in control cells. Polyak et al. (1997) [PubMed 9305847] termed these genes 'p53-induced genes,' or PIGs, several of which were predicted to encode redox-controlling proteins. They noted that reactive oxygen species (ROS) are potent inducers of apoptosis. Flow cytometric analysis showed that p53 expression induces ROS production, which increases as apoptosis progresses under some conditions. The authors stated that the PIG10 gene, also called ENC1, encodes an actin-binding protein.[supplied by OMIM]

Sequence: MSVSVHENRKSRASSGSINIYLFHKSSYADSVLTHLNLLRQQRLFTDVLLHAGNRTFPCHRAVLAACSRYFEAMFSGGLKESQDSEVNFDNSIHPEVLELLLDYAYSSRVIINEENAESLLEAGDMLEFQDIRDACAEFLEKNLHPTNCLGMLLLSDAHQCTKLYELSWRMCLSNFQTIRKNEDFLQLPQDMVVQLLSSEELETEDERLVYESAINWISYDLKKRYCYLPELLQTVRLALLPAIYLMENVAMEELITKQRKSKEIVEEAIRCKLKILQNDGVVTSLCARPRKTGHALFLLGGQTFMCDKLYLVDQKAKEIIPKADIPSPRKEFSACAIGCKVYITGGRGSENGVSKDVWVYDTLHEEWSKAAPMLVARFGHGSAELKHCLYVVGGHTAATGCLPASPSVSLKQVEHYDPTINKWTMVAPLREGVSNAAVVSAKLKLFAFGGTSVSHDKLPKVQCYDQCENRWTVPATCPQPWRYTAAAVLGNQIFIMGGDTEFSACSAYKFNSETYQWTKVGDVTAKRMSCHAVASGNKLYVVGGYFGIQRCKTLDCYDPTLDVWNSITTVPYSLIPTAFVSTWKHLPS
Specifications

Specifications

NP_003624.1
Liquid
8507
ENC1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSVSVHENRKSRASSGSINIYLFHKSSYADSVLTHLNLLRQQRLFTDVLLHAGNRTFPCHRAVLAACSRYFEAMFSGGLKESQDSEVNFDNSIHPEVLELLLDYAYSSRVIINEENAESLLEAGDMLEFQDIRDACAEFLEKNLHPTNCLGMLLLSDAHQCTKLYELSWRMCLSNFQTIRKNEDFLQLPQDMVVQLLSSEELETEDERLVYESAINWISYDLKKRYCYLPELLQTVRLALLPAIYLMENVAMEELITKQRKSKEIVEEAIRCKLKILQNDGVVTSLCARPRKTGHALFLLGGQTFMCDKLYLVDQKAKEIIPKADIPSPRKEFSACAIGCKVYITGGRGSENGVSKDVWVYDTLHEEWSKAAPMLVARFGHGSAELKHCLYVVGGHTAATGCLPASPSVSLKQVEHYDPTINKWTMVAPLREGVSNAAVVSAKLKLFAFGGTSVSHDKLPKVQCYDQCENRWTVPATCPQPWRYTAAAVLGNQIFIMGGDTEFSACSAYKFNSETYQWTKVGDVTAKRMSCHAVASGNKLYVVGGYFGIQRCKTLDCYDPTLDVWNSITTVPYSLIPTAFVSTWKHLPS
RUO
ENC1
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
92.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CCL28|ENC-1|FLJ39259|KLHL35|KLHL37|NRPB|PIG10|TP53I10
ENC1
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit