Learn More
Novus Biologicals™ Human Endostatin ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP275838
Catalog No. NBP275838
Add to cart
Description
Description
The Human Endostatin ELISA Kit (Colorimetric) measures human Endostatin in serum, plasma and other biological fluids.
Specifications
Specifications
0.16 - 10 ng/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Human
1 Kit
This kit recognizes Human COL18a1 in samples. No significant cross-reactivity or interference between Human COL18a1 and analogues was observed.
80781
0.10 ng/mL
Endostatin
COL18A1
Biological Fluids, Plasma, Serum