Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ENTPD3 Partial ORF (NP_001239, 107 a.a. - 216 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00000956Q01S

 View more versions of this product

Catalog No. 89-943-310


Add to cart

Description

Description

ENTPD3 is similar to E-type nucleotidases (NTPases). NTPases, such as CD39, mediate catabolism of extracellular nucleotides. ENTPD3 contains 4 apyrase-conserved regions which is characteristic of NTPases. [provided by RefSeq]

Sequence: QDVPRAFEECMQKVKGQVPSHLHGSTPIHLGATAGMRLLRLQNETAANEVLESIQSYFKSQPFDFRGAQIISGQEEGVYGWITANYLMGNFLEKNLWHMWVHPHGVETTG
Specifications

Specifications

NP_001239
Liquid
956
ENTPD3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD39L3|FLJ93839|HB6|NTPDase-3
ENTPD3
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QDVPRAFEECMQKVKGQVPSHLHGSTPIHLGATAGMRLLRLQNETAANEVLESIQSYFKSQPFDFRGAQIISGQEEGVYGWITANYLMGNFLEKNLWHMWVHPHGVETTG
RUO
ENTPD3
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit