Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EPB41L1 Partial ORF (NP_818932, 1 a.a. - 88 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00002036Q01S

 View more versions of this product

Catalog No. 89-943-350


Add to cart

Description

Description

Erythrocyte membrane protein band 4.1 (EPB41) is a multifunctional protein that mediates interactions between the erythrocyte cytoskeleton and the overlying plasma membrane. The protein encoded by this gene is a neuronally-enriched protein that is structurally similar to EPB41. The encoded protein binds and stabilizes D2 and D3 dopamine receptors at the neuronal plasma membrane. Multiple transcript variants encoding different isoforms have been found for this gene, but the full-length nature of only two of them has been determined. [provided by RefSeq]

Sequence: MEEKDYSEADGLSERTTPSKAQKSPQKIAKKYKSAICRVTLLDASEYECEVEKHGRGQVLFDLVCEHLNLLEKDYFGLTFCDADSQKN
Specifications

Specifications

NP_818932
Liquid
2036
EPB41L1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
4.1N|DKFZp686H17242|KIAA0338|MGC11072
EPB41L1
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.42kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MEEKDYSEADGLSERTTPSKAQKSPQKIAKKYKSAICRVTLLDASEYECEVEKHGRGQVLFDLVCEHLNLLEKDYFGLTFCDADSQKN
RUO
EPB41L1
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit