Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EPB41L5 Full-length ORF (NP_065960.2, 1 a.a. - 733 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00057669P01L

 View more versions of this product

Catalog No. 89-943-363

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Sequence: MLSFFRRTLGRRSMRKHAEKERLREAQRAATHIPAAGDSKSIITCRVSLLDGTDVSVDLPKKAKGQELFDQIMYHLDLIESDYFGLRFMDSAQVAHWLDGTKSIKKQVKIGSPYCLHLRVKFYSSEPNNLREELTRYLFVLQLKQDILSGKLDCPFDTAVQLAAYNLQAELGDYDLAEHSPELVSEFRFVPIQTEEMELAIFEKWKEYRGQTPAQAETNYLNKAKWLEMYGVDMHVVKARDGNDYSLGLTPTGVLVFEGDTKIGLFFWPKITRLDFKKNKLTLVVVEDDDQGKEQEHTFVFRLDHPKACKHLWKCAVEHHAFFRLRGPVQKSSHRSGFIRLGSRFRYSGKTEYQTTKTNKARRSTSFERRPSKRYSRRTLQMKACATKPEELSVHNNVSTQSNGSQQAWGMRSALPVSPSISSAPVPVEIENLPQSPGTDQHDRKCIPLNIDLLNSPDLLEATIGDVIGASDTMETSQALNDVNVATRLPGLGEPEVEYETLKDTSEKLKQLEMENSPLLSPRSNIDVNINSQEEVVKLTEKCLNNVIESPGLNVMRVPPDFKSNILKAQVEAVHKVTKEDSLLSHKNANVQDAATNSAVLNENNVPLPKESLETLMLITPADSGSVLKEATDELDALLASLTENLIDHTVAPQVSSTSMITPRWIVPQSGAMSNGLAGCEMLLTGKEGHGNKDGISLISPPAPFLVDAVTSSGPILAEEAVLKQKCLLTTEL
Specifications

Specifications

NP_065960.2
Liquid
57669
Human EPB41L5 Full-length ORF (NP_065960.2, 1 a.a. - 733 a.a.) Recombinant Protein with GST-tag at N-terminal
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at −80°C. Aliquot to avoid repeated freezing and thawing.
BE37, FLJ12957, KIAA1548
EPB41L5
Wheat Germ (in vitro)
GST Tag
AP, Array, ELISA, WB-Re
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
108.3kDa
Glutathione Sepharose 4 Fast Flow
25μg
RUO
EPB41L5
wheat germ expression system
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit