Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EPB42 Full-length ORF (AAH99627.1, 1 a.a. - 619 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00002038P01

Catalog No. 89-943-366


Add to cart

Description

Description

Erythrocyte membrane protein band 4.2 is an ATP-binding protein which may regulate the association of protein 3 with ankyrin. It probably has a role in erythrocyte shape and mechanical property regulation. Mutations in the EPB42 gene are associated with recessive spherocytic elliptocytosis and recessively transmitted hereditary hemolytic anemia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MGQALGIKSCDFQAARNNEEHHTKALSSRRLFVRRGQPFTIILYFRAPVRAFLPALKKVALTAQTGEQPSKINRTQATFPISSLGDRKWWSAVVEERDAQSWTISVTTPADAVIGHYSLLLQVSGRKQLLLGQFTLLFNPWNREDAVFLKNEAQRMEYLLNQNGLIYLGTADCIQAESWDFGQFEGDVIDLSLRLLSKDKQVEKWSQPVHVARVLGALLHFLKEQRVLPTPQTQATQEGALLNKRRGSVPILRQWLTGRGRPVYDGQAWVLAAVACTVLRCLGIPARVVTTFASAQGTGGRLLIDEYYNEEGLQNGEGQRGRIWKCCEDGTLELTDSNTKYVGNNISTKGVGSDRCEDITQNYKYPEGSLQEKEVLERVEKEKMEREKDNGIRPPSLETASPLYLLLKAPSSLPLRGDAQISVTLVNHSEQEKAVQLAIGVQAVHYNGVLAAKLWRKKLHLTLSANLEKIITIGLFFSNFERNPPENTFLRLTAMATHSESNLSCFAQEDIAICRPHLAIKMPEKAEQYQPLTASVSLQNSLDAPMEDCVISILGRGLIHRERSYRFRSVWPENTMCAKFQFTPTHVGLQRLTVEVDCNMFQNLTNYKSVTVVAPELSA
Specifications

Specifications

AAH99627.1
Liquid
2038
EPB42 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGQALGIKSCDFQAARNNEEHHTKALSSRRLFVRRGQPFTIILYFRAPVRAFLPALKKVALTAQTGEQPSKINRTQATFPISSLGDRKWWSAVVEERDAQSWTISVTTPADAVIGHYSLLLQVSGRKQLLLGQFTLLFNPWNREDAVFLKNEAQRMEYLLNQNGLIYLGTADCIQAESWDFGQFEGDVIDLSLRLLSKDKQVEKWSQPVHVARVLGALLHFLKEQRVLPTPQTQATQEGALLNKRRGSVPILRQWLTGRGRPVYDGQAWVLAAVACTVLRCLGIPARVVTTFASAQGTGGRLLIDEYYNEEGLQNGEGQRGRIWKCCEDGTLELTDSNTKYVGNNISTKGVGSDRCEDITQNYKYPEGSLQEKEVLERVEKEKMEREKDNGIRPPSLETASPLYLLLKAPSSLPLRGDAQISVTLVNHSEQEKAVQLAIGVQAVHYNGVLAAKLWRKKLHLTLSANLEKIITIGLFFSNFERNPPENTFLRLTAMATHSESNLSCFAQEDIAICRPHLAIKMPEKAEQYQPLTASVSLQNSLDAPMEDCVISILGRGLIHRERSYRFRSVWPENTMCAKFQFTPTHVGLQRLTVEVDCNMFQNLTNYKSVTVVAPELSA
RUO
EPB42
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
95.9kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC116735|MGC116737|PA
EPB42
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit