Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EPB49 Full-length ORF (AAH52805.1, 1 a.a. - 383 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00002039P01L

 View more versions of this product

Catalog No. 89-943-369


Add to cart

Description

Description

Dematin, or EPB49, is an actin-bundling protein originally identified in the erythroid membrane skeleton. Its actin-bundling activity is abolished upon phosphorylation by cAMP-dependent protein kinase and is restored after dephosphorylation (Rana et al., 1993 [PubMed 8341682]).[supplied by OMIM]

Sequence: MERLQKQPLTSPGSVSPSRDSSVPGSPSSIVAKMDNQVLGYKDLAAIPKDKAILDIERPDLMIYEPHFTYSLLEHVELPRSRERSLSPKSTSPPPSPEVWADSRSPGIISQASAPRTTGTPRTSLPHFHHPETSRPDSNIYKKPPIYKQRESVGGSPQTKHLIEDLIIESSKFPAAQPPDPNQPAKIETDYWPCPPSLAVVETEWRKRKASRRGAEEEEEEEDDDSGEEMKALRERQREELSKVTSNLGKMILKEEMEKSLPIRRKTRSLPDRTPFHTSLHQGTSKSSSLPAYGRTTLSRLQSTEFSPSGSETGSPGLQIYPYEMLVVTNKGRTKLPPGVDRMRLERHLSAEDFSRVFAMSPEEFGKLALWKRNELKKKASLF
Specifications

Specifications

AAH52805.1
Liquid
2039
EPB49 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MERLQKQPLTSPGSVSPSRDSSVPGSPSSIVAKMDNQVLGYKDLAAIPKDKAILDIERPDLMIYEPHFTYSLLEHVELPRSRERSLSPKSTSPPPSPEVWADSRSPGIISQASAPRTTGTPRTSLPHFHHPETSRPDSNIYKKPPIYKQRESVGGSPQTKHLIEDLIIESSKFPAAQPPDPNQPAKIETDYWPCPPSLAVVETEWRKRKASRRGAEEEEEEEDDDSGEEMKALRERQREELSKVTSNLGKMILKEEMEKSLPIRRKTRSLPDRTPFHTSLHQGTSKSSSLPAYGRTTLSRLQSTEFSPSGSETGSPGLQIYPYEMLVVTNKGRTKLPPGVDRMRLERHLSAEDFSRVFAMSPEEFGKLALWKRNELKKKASLF
RUO
EPB49
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
69.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DMT|FLJ78462|FLJ98848
EPB49
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit