Learn More
R&D Systems™ Human EpCAM/TROP1 His-tag Recombinant Protein, Carrier-Free Available on GSA/VA Contract for Federal Government customers only.
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Manufacturer:  R&D Systems™ 9277EP050
Catalog No. 92-77E-P050
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human EpCAM/TROP1 His-tag Protein is derived from HEK293. The Recombinant Human EpCAM/TROP1 His-tag Protein has been validated for the following applications: Bioactivity.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
The product is shipped at ambient temperature.
Specifications
Specifications
P16422
Lyophilized from a 0.2μm filtered solution in PBS and NaCl
30 to 42 kDa, reducing conditions
>90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Human embryonic kidney cell, HEK293-derived
RUO
0.4 to 2.4μg/mL (ED 50)
Unconjugated
Yes
Bioactivity
4072
17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
<0.10 EU per 1μg of protein by LAL method
Recombinant protein
Human
Yes (His tagged)