Learn More
Abnova™ Human EPHA1 (NP_005223.3, 586 a.a. - 976 a.a.) Partial Recombinant Protein with GST-tag expressed in baculovirus infected Sf21 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P5545
Catalog No. 89-943-383
Add to cart
Description
Description
This gene belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. This gene is expressed in some human cancer cell lines and has been implicated in carcinogenesis. [provided by RefSeq]
Specifications
Specifications
NP_005223.3
Liquid
2041
EPHA1 (Human) Recombinant Protein
Glutathione sepharose chromatography
Loading 1 ug protein in SDS-PAGE
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EPH|EPHT|EPHT1|MGC163163
EPHA1
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg(or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip 3000. Substrate: Blk/Lyntide. ATP: 100 uM.
Yes
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
72kDa
Baculovirus infected insect cell (Sf21) expression system
82% by SDS-PAGE/CBB staining
5 ug
RUO
EPHA1
Baculovirus infected insect cell (Sf21) expression system
Insect
GST