Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EPHA10 Full-length ORF (AAH67734.1, 1 a.a. - 295 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00284656P01S

 View more versions of this product

Catalog No. 89-943-386


Add to cart

Description

Description

Ephrin receptors, the largest subfamily of receptor tyrosine kinases (RTKs), and their ephrin ligands are important mediators of cell-cell communication regulating cell attachment, shape, and mobility in neuronal and epithelial cells (Aasheim et al., 2005 [PubMed 15777695]). See MIM 179610 for additional background on Eph receptors and ephrins.[supplied by OMIM]

Sequence: METCAGPHPLRLFLCRMQLCLALLLGPWRPGTAEEVILLDSKASQAELGWTALPSNGWEEISGVDEHDRPIRTYQVCNVLEPNQDNWLQTGWISRGRGQRIFVELQFTLRDCSSIPGAAGTCKETFNVYYLETEADLGRGRPRLGGSRPRKIDTIAADESFTQGDLGERKMKLNTEVREIGPLSRRGFHLAFQDVGACVALVSVRVYYKQCRATVRGLATFPATAAESAFSTLVEVAGTCVAHSEGEPGSPPRMHCGADGEWLVPVGRCSCSAGFQERGDFCEGIQLAGGRGVGV
Specifications

Specifications

AAH67734.1
Liquid
284656
EPHA10 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
METCAGPHPLRLFLCRMQLCLALLLGPWRPGTAEEVILLDSKASQAELGWTALPSNGWEEISGVDEHDRPIRTYQVCNVLEPNQDNWLQTGWISRGRGQRIFVELQFTLRDCSSIPGAAGTCKETFNVYYLETEADLGRGRPRLGGSRPRKIDTIAADESFTQGDLGERKMKLNTEVREIGPLSRRGFHLAFQDVGACVALVSVRVYYKQCRATVRGLATFPATAAESAFSTLVEVAGTCVAHSEGEPGSPPRMHCGADGEWLVPVGRCSCSAGFQERGDFCEGIQLAGGRGVGV
RUO
EPHA10
wheat germ expression system
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
58.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ16103|FLJ33655|MGC43817
EPHA10
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit