Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human EPHA10 Full-length ORF (AAH67734.1, 1 a.a. - 295 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00284656P01L
Description
Ephrin receptors, the largest subfamily of receptor tyrosine kinases (RTKs), and their ephrin ligands are important mediators of cell-cell communication regulating cell attachment, shape, and mobility in neuronal and epithelial cells (Aasheim et al., 2005 [PubMed 15777695]). See MIM 179610 for additional background on Eph receptors and ephrins.[supplied by OMIM]
Sequence: METCAGPHPLRLFLCRMQLCLALLLGPWRPGTAEEVILLDSKASQAELGWTALPSNGWEEISGVDEHDRPIRTYQVCNVLEPNQDNWLQTGWISRGRGQRIFVELQFTLRDCSSIPGAAGTCKETFNVYYLETEADLGRGRPRLGGSRPRKIDTIAADESFTQGDLGERKMKLNTEVREIGPLSRRGFHLAFQDVGACVALVSVRVYYKQCRATVRGLATFPATAAESAFSTLVEVAGTCVAHSEGEPGSPPRMHCGADGEWLVPVGRCSCSAGFQERGDFCEGIQLAGGRGVGVSpecifications
AAH67734.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
58.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ16103/FLJ33655/MGC43817 | |
EPHA10 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
284656 | |
EPHA10 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
METCAGPHPLRLFLCRMQLCLALLLGPWRPGTAEEVILLDSKASQAELGWTALPSNGWEEISGVDEHDRPIRTYQVCNVLEPNQDNWLQTGWISRGRGQRIFVELQFTLRDCSSIPGAAGTCKETFNVYYLETEADLGRGRPRLGGSRPRKIDTIAADESFTQGDLGERKMKLNTEVREIGPLSRRGFHLAFQDVGACVALVSVRVYYKQCRATVRGLATFPATAAESAFSTLVEVAGTCVAHSEGEPGSPPRMHCGADGEWLVPVGRCSCSAGFQERGDFCEGIQLAGGRGVGV | |
RUO | |
EPHA10 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction